Arachidyl arachidate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

452020100MG
100296-386EA 303.38 USD
100296-386
Arachidyl arachidate ≥99%
Arachidyl arachidate
MDL Number: MFCD00056268
CAS Number: 22432-80-0
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR