1-Hexacosanol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

402600100MG
100296-384EA 564.92 USD
100296-384
1-Hexacosanol ≥99%
1-Hexacosanol

Store in a cool, dry place

Formula: H₃C(CH₂)₂₄CH₂OH
MDL Number: MFCD00002940
CAS Number: 506-52-5
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a832ca77da0c664749c5bb3b398e2c095831d97&vid=6a832ca77da0c664749c5bb3b398e2c095831d97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR