Trierucin (13C) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

332210100MG
100296-378EA 341.31 USD
100296-378
Trierucin (13C) ≥99%
Trierucin (13C)
MDL Number: MFCD00056321
CAS Number: 2752-99-0
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46433f2561be7197b75e9da4f5ec3e9e4e3dd0f&vid=f46433f2561be7197b75e9da4f5ec3e9e4e3dd0f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR