Tri-γ-linolenin (6C,9C,12C) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

331835100MG
100296-374EA 549.23 USD
100296-374
Tri-γ-linolenin (6C,9C,12C) ≥99%
Trigammalinolenin (6C,9C,12C)

Store in a cool, dry place

MW: 873.50 g/mol
MDL Number: MFCD02259104
UN: 12352100
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR