Trilinolenin (9C,12C,15C) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

331830100MG
100296-372EA 384.46 USD
100296-372
Trilinolenin (9C,12C,15C) ≥99%
Trilinolenin (9C,12C,15C)
MW: 873.50 g/mol
MDL Number: MFCD00042898
CAS Number: 14465-68-0
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR