Dilinoelaidin (9Tr,12Tr) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

32186025MG
100296-366EA 341.31 USD
100296-366
Dilinoelaidin (9Tr,12Tr) ≥99%
Dilinoelaidin (9Tr,12Tr)

Store in a cool, dry place

MW: 616.90 g/mol
MDL Number: MFCD00673506
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09dd8eea47813889d300c3d9f46ea86696dfb2fe&vid=2f3d0918f484a3a35734b8bafdb1b4f3d641d6f2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR