3-Hydroxyhexadecanoic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

14160350MG
100295-796EA 614.62 USD
100295-796
3-Hydroxyhexadecanoic acid ≥98%
3-Hydroxyhexadecanoic acid
Formula: C₁₆H₃₂O₃
MW: 272.42 g/mol
MDL Number: MFCD00171638
CAS Number: 2398-34-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78397c96e3f180253290e220186bea51f63b3bba&vid=78397c96e3f180253290e220186bea51f63b3bba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR