9(Z)-Tetradecanoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

101401100MG
100295-772EA 528.31 USD
100295-772
9(Z)-Tetradecanoic acid ≥99%
9(Z)-Tetradecanoic acid
MW: 226.38 g/mol
MDL Number: MFCD00004436
CAS Number: 544-64-9
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR