1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

0940031GM
100295-710EA 243.23 USD
100295-710
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one ≥97%
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one
Formula: C₁₁H₉F₃O
MDL Number: MFCD00052843
CAS Number: 80992-92-3
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5168580c99ff559569492e58ccb4d9a03223c823&vid=5168580c99ff559569492e58ccb4d9a03223c823&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR