2,3,4,5-Tetrafluorobenzamide

Supplier: INDOFINE CHEMICAL COMPANY MS

075225GM
100295-438EA 258.92 USD
100295-438
2,3,4,5-Tetrafluorobenzamide
2,3,4,5-Tetrafluorobenzamide
Formula: C₇H₃F₄NO
MW: 193.1
MDL Number: MFCD01631457
CAS Number: 16582-94-8
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a032033cc690006e8f34df52496d2b908464193d&vid=a032033cc690006e8f34df52496d2b908464193d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR