Boc-D-Cha-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-β-cyclohexyl-D-Ala-OH

046171GM
100295-184EA 115.08 USD
100295-184
Boc-D-Cha-OH ≥99%
Boc-D-Cha-OH
Formula: C₁₄H₂₅NO₄
MW: 271.357 g/mol
MDL Number: MFCD00133627
CAS Number: 127095-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR