Boc-L-3-benzothienylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-β-(3-benzothienyl)-Ala-OH, (S)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

045871GM
100295-172EA 196.15 USD
100295-172
Boc-L-3-benzothienylalanine ≥99%
Boc-L-3-benzothienylalanine
Formula: C₁₆H₁₉NO₄S
MDL Number: MFCD00237538
CAS Number: 154902-51-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a38f065a69b5e2204485704690018778cce39c6e&vid=cbee27acc2fd21fc1aab9eaafc8f47ae6507bf94&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2134a85fc4429667d9bb3d862ffff3f58cf54717&vid=3d0704ecee8f74d7697a63c6e121d707668c611f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR