1-Boc-4-(aminomethyl)piperidine ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

045461GM
100295-142EA 31.38 USD
100295-142
1-Boc-4-(aminomethyl)piperidine ≥95%
1-Boc-4-(aminomethyl)piperidine

  • Light yellow oil or low melting point crystalline mass
  • Store at 0-8 °C

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD01076207
CAS Number: 144222-22-0
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc2091acf56616c0cd1221483929ed223f6cbea&vid=5fc2091acf56616c0cd1221483929ed223f6cbea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR