Mono(boc)(PEO)-3-diamine, boc-DOOA ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-(tert-Butoxycarbonyl)-2,2'-(ethylenedioxy)diethylamine, N-Boc-3,6-dioxa-1,8-octanediamine, tert-Butyl [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamate, [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamic acid tert-Butyl ester, N-Boc-2,2'-(ethylenedioxy)diethylamine, N-(tert-Butoxycarbonyl)-3,6-dioxa-1,8-octanediamine, tert-Butyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate

045411GM
100295-140EA 338.69 USD
100295-140
Mono(boc)(PEO)-3-diamine, boc-DOOA ≥98%
Mono(boc)(PEO)-3-diamine, boc-DOOA

  • Yellow oil
  • Store at 0-8 °C

Formula: C₁₁H₂₄N₂O₄
MW: 248.3 g/mol
MDL Number: MFCD03788155
CAS Number: 153086-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1874639115fe2caaafa32824c2558604c3d24e5&vid=a1874639115fe2caaafa32824c2558604c3d24e5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e506a3acda5547795ba75fca5a8edf67354c5c9c&vid=e506a3acda5547795ba75fca5a8edf67354c5c9c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR