L-α-Aminoadipic acid ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid

042431GM
100294-918EA 124.23 USD
100294-918
L-α-Aminoadipic acid ≥98%, powder
L-α-Aminoadipic acid

  • White powder
  • Store in a cool, dry place

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00002636
CAS Number: 1118-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94ca1df412f777627b98e255c25a958cb134972c&vid=94ca1df412f777627b98e255c25a958cb134972c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR