Fmoc-3-(1-naphthyl)-L-alanine ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-1-Nal-OH

0413331GM
100294-580EA 128.15 USD
100294-580
Fmoc-3-(1-naphthyl)-L-alanine ≥99%, powder
Fmoc-3-(1-naphthyl)-L-alanine

  • White powder
  • Store at 0-8 °C

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Melting Pt: 178…188 °C
MDL Number: MFCD00151914
CAS Number: 96402-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4361d5ee0284654f0084025f0ce0582d1919b3b&vid=d8a57fc7fd9c1f1f384e1fc88fb50af0f4e86474&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR