Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

04047100GM
100294-430EA 300.77 USD
100294-430
Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98% (by HPLC), crystalline powder
Ethyl N-acetylglycinate (Ac-Gly-OEt)

  • White or off-white crystalline powder
  • Store at 0-8 °C

Formula: C₆H₁₁NO₃
Melting Pt: 42…48 °C
MDL Number: MFCD00009173
CAS Number: 1906-82-7
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39d527f76fd29eaf97923aa145a584a14ccef0d8&vid=39d527f76fd29eaf97923aa145a584a14ccef0d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR