3-Fluoro-p-anisaldehyde ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-Fluoro-4-methoxybenzaldehyde, 3-Fluoro-4-anisaldehyde

269575GM
100294-240EA 104.62 USD
100294-240
3-Fluoro-p-anisaldehyde ≥98%
3-Fluoro-p-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14
Melting Pt: 34…35 °C
MDL Number: MFCD00003349
CAS Number: 351-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6be5803e370ab5b65b98055fb4aeeb273c175501&vid=6be5803e370ab5b65b98055fb4aeeb273c175501&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR