Hexadecafluorosebaconitrile ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Perfluorosebaconitrile

2616181GM
100293-782EA 187 USD
100293-782
Hexadecafluorosebaconitrile ≥97%
Hexadecafluorosebaconitrile
Formula: C₁₀F₁₆N₂
MW: 452.09 g/mol
Boiling Pt: 147…148 °C
MDL Number: MFCD00042366
CAS Number: 2342-09-8
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR