Methyl-3-hydroxyundecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

24110350MG
100293-676EA 588.46 USD
100293-676
Methyl-3-hydroxyundecanoate ≥98%
Methyl-3-hydroxyundecanoate

Store in a cool, dry place

Formula: C₁₂H₂₄O₃
MDL Number: MFCD00673454
CAS Number: 129758-71-0
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a714c3afc1cc44aea87523795d99606a7f24d1e&vid=3a714c3afc1cc44aea87523795d99606a7f24d1e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR