3-Fluoro-o-xylene ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Fluoro-2,3-dimethylbenzene, 1,2-Dimethyl-3-fluorobenzene

0972025GM
100293-532EA 120.31 USD
100293-532
3-Fluoro-o-xylene ≥98%
3-Fluoro-o-xylene
Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 148…152 °C
MDL Number: MFCD00000323
CAS Number: 443-82-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR