N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Bromobenzyl Succinimidyl Carbonate, Carbonic Acid 2-Bromobenzyl Succinimidyl Ester, Z(2-Br)-OSu

0494925GM
100293-026EA 105.92 USD
100293-026
N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%
N-(2-Bromobenzyloxycarbonyloxy)succinimide

  • White Solid
  • Store at 0-8 °C

Formula: C₁₂H₁₀BrNO₅
MW: 328.12 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD00082848
CAS Number: 128611-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=154278a903d5652655d364e0e65f63fb0bddcdb0&vid=154278a903d5652655d364e0e65f63fb0bddcdb0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR