L-Aspartic acid 4-cyclohexyl ester ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Asp(OcHex)-OH hydrochloride salt, H-Asp(OcHx)-OH, H-Asp(OcHex)-OH

043695GM
100292-828EA 115.08 USD
100292-828
L-Aspartic acid 4-cyclohexyl ester ≥98%, powder
L-Aspartic acid 4-cyclohexyl ester

  • White to off white powder
  • Store in a cool, dry place

Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
MDL Number: MFCD00190762
CAS Number: 112259-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4698bf82e6a5bee332b35b95c2c5839993ceb5e8&vid=4698bf82e6a5bee332b35b95c2c5839993ceb5e8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f10a29498deb98686e4d8eadfef4cfd319c696d&vid=6f10a29498deb98686e4d8eadfef4cfd319c696d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR