N(α)-Cbz-D-tryptophan ≥98% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

0425705GM
100292-722EA 105.92 USD
100292-722
N(α)-Cbz-D-tryptophan ≥98% (by HPLC)
N(α)-Cbz-D-tryptophan
Formula: C₁₉H₁₈N₂O₄
Melting Pt: 118…124 °C
MDL Number: MFCD00037945
CAS Number: 2279-15-4
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c24adcbb10dc1b0c90b00e8e6f57af93acf8ed9&vid=2e641e63656b0bed0923e200fd6b222ebf6b24cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR