Tris(carbobenzoxy)-L-arginine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nα,Nδ,Nomega-Tris(benzyloxycarbonyl)-L-arginine, Tri-Cbz-L-arginine, Z-Arg(Z)2-OH, Z-Arg(Cbz)₂-OH

0425405GM
100292-706EA 281.15 USD
100292-706
Tris(carbobenzoxy)-L-arginine ≥99%
Tris(carbobenzoxy)-L-arginine
Formula: C₃₀H₃₂N₄O₈
MW: 576.61 g/mol
Melting Pt: 137…140 °C
MDL Number: MFCD00038693
CAS Number: 14611-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR