(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Lys(Z)-OBzl.HCl, H-Lys(Z)-OBzl·HCl, Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride

0420175GM
100292-490EA 151.69 USD
100292-490
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥99%, crystalline powder
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₂₁H₂₆N₂O₄·HCl
Melting Pt: 136…141 °C
MDL Number: MFCD00038981
CAS Number: 6366-70-7
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe5da90c835277d179cf19df8e586ca4959d6bdc&vid=817dd5c5ba4b4629fd78e41d65fa5a39cf15653f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR