Dodecanoic-D23 acid ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Lauric-d23 acid

871223100MG
100291-974EA 494.31 USD
100291-974
Dodecanoic-D23 acid ≥95%
Dodecanoic-D23 acid

Store in a cool, dry place

Formula: C₁₂H₂₄O₂
MW: 223.50 g/mol
MDL Number: MFCD00144567
CAS Number: 59154-43-7
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR