Octadecanoic Acid-1-13C (Stearic) ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Heptadecanecarboxylic acid-1-13C

781800100MG
100291-958EA 457.69 USD
100291-958
Octadecanoic Acid-1-13C (Stearic) ≥98%
Octadecanoic Acid-1-13C (Stearic)

Store in a cool, dry place

Formula: CH₃(CH₂)₁₆₁₃CO₂H
MW: 285.47 g/mol
MDL Number: MFCD00002753
CAS Number: 85541-42-0
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR