Hexadecanoic acid-2-¹³C (Palmitic) ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

781602100MG
100291-950EA 978.15 USD
100291-950
Hexadecanoic acid-2-¹³C (Palmitic) ≥98%
Hexadecanoic acid-2-13C (Palmitic)

Store in a cool, dry place

Formula: CH₃(CH₂)₁₃¹³CH₂CO₂H
MDL Number: MFCD00083983
CAS Number: 287100-87-2
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR