Octadecanoic-2,2-D2 acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Stearic acid-2,2-d2

711802100MG
100291-836EA 398.85 USD
100291-836
Octadecanoic-2,2-D2 acid ≥99%
Octadecanoic-2,2-D2 acid

Store in a cool, dry place

Formula: CH₃(CH₂)₁₅CD₂CO₂H
MW: 286.50 g/mol
MDL Number: MFCD00053340
CAS Number: 19905-58-9
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d7edd79836650d83d27c5ebfca2554e34e39dad&vid=6d7edd79836650d83d27c5ebfca2554e34e39dad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR