Isobutyryl (D7)-L-carnitine.Hcl

Supplier: INDOFINE CHEMICAL COMPANY MS

71177410MG
100291-820EA 1093.23 USD
100291-820
Isobutyryl (D7)-L-carnitine.Hcl
Isobutyryl (D7)-L-carnitine.Hcl

Store in a cool, dry place

MDL Number: MFCD05864272
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR