Acetyl-(D3)-L-carnitine.HCl

Supplier: INDOFINE CHEMICAL COMPANY MS

71173210MG
100291-768EA 895.77 USD
100291-768
Acetyl-(D3)-L-carnitine.HCl
Acetyl-(D3)-L-carnitine.HCl
MDL Number: MFCD06656338
CAS Number: 362049-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67b750c9edcded0f00f42431aa5a39ec9b459a0e&vid=67b750c9edcded0f00f42431aa5a39ec9b459a0e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR