Hexadecanoic-2,2-D2 acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

711602100MG
100291-740EA 405.38 USD
100291-740
Hexadecanoic-2,2-D2 acid ≥99%
Hexadecanoic-2,2-D2 acid
MW: 258.40 g/mol
MDL Number: MFCD00053337
CAS Number: 62689-96-7
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR