Sodium glycocholate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glycocholic acid, sodium salt hydrate, Sodium ((4R)-4-((3R,7R,10S,12S,13R,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoyl)glycinate, Glycocholic acid sodium

6510081GM
100291-584EA 115.08 USD
100291-584
Sodium glycocholate ≥98%
Sodium glycocholate

Store in a cool, dry place

Formula: C₂₆H₄₂NNaO₆
MW: 487.60 g/mol
MDL Number: MFCD00036741
CAS Number: 863-57-0
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aefa292474e2bc967f5b9a8ea378cb1a0e54b599&vid=aefa292474e2bc967f5b9a8ea378cb1a0e54b599&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR