Methyl cholate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

651002100MG
100291-574EA 341.31 USD
100291-574
Methyl cholate ≥98%
Methyl cholate
Formula: C₂₅H₄₂O₅
MDL Number: MFCD00064934
CAS Number: 1448-36-8
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75cfd79f0b131c6696583987f707732853309536&vid=75cfd79f0b131c6696583987f707732853309536&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR