C20-D-erythro-sphingosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

5613255MG
100291-444EA 1209.62 USD
100291-444
C20-D-erythro-sphingosine ≥98%
C20-D-erythro-sphingosine

Store in a cool, dry place

Formula: C₂₉H₄₁NO₂
MW: 327.40 g/mol
MDL Number: MFCD02259131
CAS Number: 6918-49-6
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0215d4bc2fffc07ac718fa36efa73fe55678d3c1&vid=0215d4bc2fffc07ac718fa36efa73fe55678d3c1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR