Glucopsychosine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Lyso-Ceramideglucoside

5611415MG
100291-396EA 1108.92 USD
100291-396
Glucopsychosine ≥99%
Glucopsychosine

Store in a cool, dry place

Formula: C₂₄H₄₇NO₇
MW: 461.64 g/mol
MDL Number: MFCD00079314
CAS Number: 52050-17-6
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bf8401f32035d62afc55f37d477b89fa1e1aa18&vid=0bf8401f32035d62afc55f37d477b89fa1e1aa18&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR