N-Myristoyl-D-sphingosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: C14 Ceramide

5611215MG
100291-388EA 320.38 USD
100291-388
N-Myristoyl-D-sphingosine ≥98%
N-Myristoyl-D-sphingosine

Store in a cool, dry place

Formula: C₃₂H₆₃NO₃
MDL Number: MFCD01320386
CAS Number: 34227-72-0
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68bf517529aa646685c420133c328aa028f0b50c&vid=68bf517529aa646685c420133c328aa028f0b50c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR