Disialoganglioside Gd1B ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Ganglioside GD1b disialo, GD1b

5611041MG
100291-376EA 1292 USD
100291-376
Disialoganglioside Gd1B ≥98%
Disialoganglioside Gd1B

Store in a cool, dry place

Formula: C₈₄H₁₄₆N₄O₃₉
MDL Number: MFCD00466941
CAS Number: 19553-76-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=196969209525207c3983ac1b20e9542277b30931&vid=b7f65688adc7d3cca95ddf79dc5d34386ab8c109&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR