D-erythro-Sphingosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

56108610MG
100291-344EA 490.38 USD
100291-344
D-erythro-Sphingosine ≥98%
D-erythro-Sphingosine

Store in a cool, dry place


Store in a cool, dry place

Formula: C₁₈H₃₇NO₂
MW: 299.50 g/mol
MDL Number: MFCD00036751
CAS Number: 123-78-4
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22453b85568218f9e05e49b1dfaa50f6e595c4a&vid=a22453b85568218f9e05e49b1dfaa50f6e595c4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR