N-Hexanoylsphingosylphosphorylcholine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

5610835MG
100291-336EA 1065.77 USD
100291-336
N-Hexanoylsphingosylphosphorylcholine ≥98%
N-Hexanoylsphingosylphosphorylcholine

Store in a cool, dry place

MDL Number: MFCD01073551
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44a9b29c142fc1c9d15de352f3a432b10e83133a&vid=54f91aa8efe7bfbb46a3392b5e9132aebd73f546&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR