Globosides (Gl4, Gb4, Globotetraosyl Ceramide) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

5610655MG
100291-302EA 1909.23 USD
100291-302
Globosides (Gl4, Gb4, Globotetraosyl Ceramide) ≥99%
Globosides (Gl4, Gb4, Globotetraosyl Ceramide)

Store in a cool, dry place

MW: 1239 g/mol (C18:0)
MDL Number: MFCD00131163
UN: 12352100
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR