1-Hexadecanol (cetyl alcohol) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

4016001GM
100291-076EA 189.62 USD
100291-076
1-Hexadecanol (cetyl alcohol) ≥99%
1-Hexadecanol

Storage conditions: Store in a cool, dry place

Formula: H₃C(CH₂)₁₄CH₂OH
MDL Number: MFCD00004760
CAS Number: 36653-82-4
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d97fb105304ba55f68c03c3e42810fa56c0f87&vid=40d97fb105304ba55f68c03c3e42810fa56c0f87&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR