NBD C6-Sphingomyelin

Supplier: INDOFINE CHEMICAL COMPANY MS

395064100UG
100291-050EA 712.69 USD
100291-050
NBD C6-Sphingomyelin
NBD C6-Sphingomyelin
Formula: C₃₅H₆₁N₆O₉P
CAS Number: 94885-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d1dd0cacb612a988e608539106315da06ee1807&vid=2d1dd0cacb612a988e608539106315da06ee1807&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR