1,2-Dihexanoyl-sn-glycero-3-phosphatidylcholine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Dihexanoyllecithin

37060025MG
100290-672EA 477.31 USD
100290-672
1,2-Dihexanoyl-sn-glycero-3-phosphatidylcholine ≥99%
1,2-Dihexanoyl-sn-glycero-3-phosphatidylcholine

Store in a cool, dry place

Formula: C₂₀H₄₀NO₈P
MW: 453.50 g/mol
MDL Number: MFCD00050431
CAS Number: 34506-67-7
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR