L-α-Phosphatidylcholines, Pc (soybean) ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

370102100MG
100290-574EA 300.77 USD
100290-574
L-α-Phosphatidylcholines, Pc (soybean) ≥98%
L-alpha-Phosphatidylcholines, Pc (soybean)

Store in a cool, dry place

MDL Number: MFCD02265636
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR