1,3-Olein-2-Stearin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1,3-di(octadec-9-enoyloxy)propan-2-yl octadecanoate

34183225MG
100290-548EA 555.77 USD
100290-548
1,3-Olein-2-Stearin ≥99%
1,3-Olein-2-Stearin

Store in a cool, dry place

Formula: C₅₇H₁₀₆O₆
MW: 887.50 g/mol
MDL Number: MFCD00272929
CAS Number: 2410-29-9
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60ee5d7403b49f2caceedac882883f98186544bf&vid=60ee5d7403b49f2caceedac882883f98186544bf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR