Tribehenin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glycerol Tridocosanoate, Glyceryl tribehenate, Glyceryl behenate, 1,2,3-Tridocosanoyl Glycerol

332200100MG
100290-468EA 302.08 USD
100290-468
Tribehenin ≥99%
Tribehenin
Formula: C₆₉H₁₃₄O₆
MW: 1059.83 g/mol
MDL Number: MFCD00056274
CAS Number: 18641-57-1
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f2a2802254740c83096d629cdd4f324eb606f28&vid=2f2a2802254740c83096d629cdd4f324eb606f28&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR