Tristearin (glycerol tristearate) ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Propane-1,2,3-triyl tristearate, Tristearin

331800100MG
100290-432EA 197.46 USD
100290-432
Tristearin (glycerol tristearate) ≥99%
Tristearin (Glycerol tristearate)
Formula: C₅₇H₁₁₀O₆
MW: 891.4797 g/mol
MDL Number: MFCD00036230
CAS Number: 555-43-1
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75d96fe081996663671af2fbc90a76372577e387&vid=75d96fe081996663671af2fbc90a76372577e387&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR