1,3-Dibehenin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

32220350MG
100290-400EA 444.62 USD
100290-400
1,3-Dibehenin ≥99%
1,3-Dibehenin

Store in a cool, dry place

MDL Number: MFCD00673516
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c190b0878c47d07efb15cd7609da63291631a15&vid=860b5c7a4d5f939a4c0bdd265db46c082cbe938c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR